MilliporeSigma
  • Infrared Ion Spectroscopy of Environmental Organic Mixtures: Probing the Composition of α-Pinene Secondary Organic Aerosol.
Infrared Ion Spectroscopy of Environmental Organic Mixtures: Probing the Composition of α-Pinene Secondary Organic Aerosol.
Environmental science & technology (2019-06-12)
Emma Q Walhout, Shelby E Dorn, Jonathan Martens, Giel Berden, Jos Oomens, Paul H-Y Cheong, Jesse H Kroll, Rachel E O'Brien
ABSTRACT
Characterizing the chemical composition of organic aerosols can elucidate aging mechanisms as well as the chemical and physical properties of the aerosol. However, the high chemical complexity and often low atmospheric abundance present a difficult analytical challenge. Milligrams or more of material may be needed for speciated spectroscopic analysis. In contrast, mass spectrometry provides a very sensitive platform but limited structural information. Here, we combine the strengths of mass spectrometry and infrared (IR) action spectroscopy to generate characteristic IR spectra of individual, mass-isolated ion populations. Soft ionization combined with in situ infrared ion spectroscopy, using the tunable free-electron laser FELIX, provides detailed information on molecular structures and functional groups. We apply this technique, along with quantum mechanical modeling, to characterize organic molecules in secondary organic aerosol (SOA) formed from the ozonolysis of α-pinene. Spectral overlap with a standard is used to identify cis-pinonic acid. We also demonstrate the characterization of isomers for multiple SOA products using both quantum mechanical computations and analyses of fragment ion spectra. These results demonstrate the detailed structural information on isolated ions obtained by combining mass spectrometry with fingerprint IR spectroscopy.