MilliporeSigma
  • Increased Dynamin-Related Protein 1-Dependent Mitochondrial Fission Contributes to High-Fat-Diet-Induced Cardiac Dysfunction and Insulin Resistance by Elevating Tafazzin in Mouse Hearts.
Increased Dynamin-Related Protein 1-Dependent Mitochondrial Fission Contributes to High-Fat-Diet-Induced Cardiac Dysfunction and Insulin Resistance by Elevating Tafazzin in Mouse Hearts.
Molecular nutrition & food research (2019-01-12)
Wenguang Chang, Dandan Xiao, Xiang Ao, Mengyang Li, Tao Xu, Jianxun Wang
ABSTRACT
High fat (HF)-diet-induced insulin resistance is a major contributor to the pathogenesis of cardiovascular diseases. However, the molecular mechanisms that regulate cardiac insulin signaling are not fully understood. The regulatory role of tafazzin in the hearts of HF-diet-fed mice is investigated. Mice are fed a HF diet or low fat (LF) diet for up to 24 weeks. After 24 weeks, it is found that HF-diet-induced cardiac dysfunction is linked to overexpression of the mitochondrial protein tafazzin. Increased tafazzin promotes mitochondrial fission and impairs insulin signaling, which is mediated by dynamin-related protein 1 (Drp-1) translocation from the cytosol to the mitochondria. Furthermore, knockdown of tafazzin with siRNA inhibits palmitic-acid-induced mitochondrial fission and restores insulin sensitivity. Moreover, miR-125b-5p as an upstream regulator targeting tafazzin is identified and palmitate-induced insulin resistance further rescued. In HF-diet-fed mouse hearts, increased tafazzin contributes to insulin resistance via mediating Drp-1 translocation to the mitochondria, and a small non-coding RNA, miR-125b-5p, at least partially regulates this signaling pathway and alleviates insulin resistance.