MilliporeSigma
  • Functional assessment of rat pulmonary flavin-containing monooxygenase activity.
Functional assessment of rat pulmonary flavin-containing monooxygenase activity.
Xenobiotica; the fate of foreign compounds in biological systems (2018-04-26)
Yildiz Yilmaz, Gareth Williams, Nenad Manevski, Markus Walles, Stephan Krähenbühl, Gian Camenisch
ABSTRACT
The expression of flavin-containing monooxygenase (FMO) varies extensively between human and commonly used preclinical species such as rat and mouse. The aim of this study was to investigate the pulmonary FMO activity in rat using benzydamine. Furthermore, the contribution of rat lung to the clearance of benzydamine was investigated using an in vivo pulmonary extraction model. Benzydamine N-oxygenation was observed in lung microsomes and lung slices. Thermal inactivation of FMO and CYP inhibition suggested that rat pulmonary N-oxygenation is predominantly FMO mediated while any contribution from CYPs is negligible. The predicted lung clearance (CLlung) estimated from microsomes and slices was 16 ± 0.6 and 2.1 ± 0.3 mL/min/kg, respectively. The results from in vivo pulmonary extraction indicated no pulmonary extraction following intravenous and intra-arterial dosing to rats. Interestingly, the predicted CLlung using rat lung microsomes corresponded to approximately 35% of rat CLliver suggesting that the lung makes a smaller contribution to the whole body clearance of benzydamine. Although benzydamine clearance in rat appears to be predominantly mediated by hepatic metabolism, the data suggest that the lung may also make a smaller contribution to its whole body clearance.