MilliporeSigma
  • Effects in Cancer Cells of the Recombinant l-Methionine Gamma-Lyase from Brevibacterium aurantiacum. Encapsulation in Human Erythrocytes for Sustained l-Methionine Elimination.
Effects in Cancer Cells of the Recombinant l-Methionine Gamma-Lyase from Brevibacterium aurantiacum. Encapsulation in Human Erythrocytes for Sustained l-Methionine Elimination.
The Journal of pharmacology and experimental therapeutics (2019-04-04)
David Machover, Luigia Rossi, Jocelyne Hamelin, Christophe Desterke, Emma Goldschmidt, Bernadette Chadefaux-Vekemans, Pascal Bonnarme, Pierre Briozzo, David Kopečný, Francesca Pierigè, Mauro Magnani, Rosella Mollicone, Farhad Haghighi-Rad, Yann Gaston-Mathé, Julien Dairou, Claude Boucheix, Raphaël Saffroy
ABSTRACT
Methionine deprivation induces growth arrest and death of cancer cells. To eliminate l-methionine we produced, purified, and characterized the recombinant pyridoxal 5'-phosphate (PLP)-dependent l-methionine γ-lyase (MGL)- BL929 from the cheese-ripening Brevibacterium aurantiacum Transformation of an Escherichia coli strain with the gene BL929 from B. aurantiacum optimized for E. coli expression led to production of the MGL-BL929. Elimination of l-methionine and cytotoxicity in vitro were assessed, and methylation-sensitive epigenetics was explored for changes resulting from exposure of cancer cells to the enzyme. A bioreactor was built by encapsulation of the protein in human erythrocytes to achieve sustained elimination of l-methionine in extracellular fluids. Catalysis was limited to α,γ-elimination of l-methionine and l-homocysteine. The enzyme had no activity on other sulfur-containing amino acids. Enzyme activity decreased in presence of serum albumin or plasma resulting from reduction of PLP availability. Elimination of l-methionine induced cytotoxicity on a vast panel of human cancer cell lines and spared normal cells. Exposure of colorectal carcinoma cells to the MGL-BL929 reduced methyl-CpG levels of hypermethylated gene promoters including that of CDKN2A, whose mRNA expression was increased, together with a decrease in global histone H3 dimethyl lysine 9. The MGL-erythrocyte bioreactor durably preserves enzyme activity in vitro and strongly eliminates l-methionine from medium.