MilliporeSigma
  • Characterization of the Key Aroma Compounds in Aged Chinese Rice Wine by Comparative Aroma Extract Dilution Analysis, Quantitative Measurements, Aroma Recombination, and Omission Studies.
Characterization of the Key Aroma Compounds in Aged Chinese Rice Wine by Comparative Aroma Extract Dilution Analysis, Quantitative Measurements, Aroma Recombination, and Omission Studies.
Journal of agricultural and food chemistry (2019-03-29)
Shuang Chen, Chengcheng Wang, Michael Qian, Zhou Li, Yan Xu
ABSTRACT
The aroma compounds in young and aged Chinese rice wines (rice wines) with a clear difference in their overall aroma profiles were analyzed by comparative aroma extract dilution analysis (cAEDA). In AEDA, more aroma-active regions with flavor dilution (FD) factors of ≥64 were detected in the aged rice wine than in the young rice wine. A total of 43 odorants were further identified and quantitated. The odor activity values (OAVs) revealed 33 aroma compounds with OAVs of ≥1 in young or aged rice wine. Among these aroma compounds with relatively higher OAVs, 3-methylbutanoic acid, 1,1-diethoxyethane, vanillin, 3-methylbutanal, sotolon, benzaldehyde, 4-vinylguaiacol, methional, and 2,3-butanedione showed significant differences between young and aged rice wines. This difference was confirmed through a quantitative analysis of 34 rice wine samples with ages of 0-15 years. Then, the aroma profile of the aged rice wine was successfully simulated through an aroma recombination model. Omission models suggested that sotolon, vanillin, 3-methylbutanal, and benzaldehyde played key roles in the overall aroma of aged rice wine.