MilliporeSigma
  • Identification of Myoferlin Expression for Prediction of Subsequent Primary Malignancy in Patients With Clear Cell Renal Cell Carcinoma.
Identification of Myoferlin Expression for Prediction of Subsequent Primary Malignancy in Patients With Clear Cell Renal Cell Carcinoma.
In vivo (Athens, Greece) (2019-07-08)
Hyun Min Koh, Hyo Jung An, Gyung Hyuck Ko, Jeong Hee Lee, Jong Sil Lee, Dong Chul Kim, Deok Ha Seo, Dae Hyun Song
ABSTRACT
Multiple primary malignant tumors are common in patients with renal cell carcinoma. However, reports on the factors that can identify patients with a risk for subsequent primary malignancies have been lacking. This study aimed to investigate whether myoferlin expression can be used as a potential marker to predict subsequent primary malignancies in patients with clear cell renal cell carcinoma (ccRCC). We evaluated the relationship of subsequent primary malignancies with clinicopathological factors and myoferlin expression in 152 patients with ccRCC, and we analyzed the strength of the association with myoferlin expression. The development of subsequent primary malignancies exhibited significant correlation with patient age (p=0.029), sex (p=0.015), T stage (p<0.001), and myoferlin expression (p=0.017). Furthermore, myoferlin hyperexpression was determined as an independent risk factor for developing a subsequent primary malignant tumor in patients with ccRCC (odds ratio(OR), 2.485, 95% Confidence Interval(CI)=1.052-5.870, p=0.038). Myoferlin hyperexpression can be a useful marker for predicting the development of subsequent primary malignancies in patients with ccRCC.