MilliporeSigma
  • Formation of α-Dicarbonyls from Dairy Related Carbohydrates with and without Nα-Acetyl-l-Lysine during Incubation at 40 and 50 °C.
Formation of α-Dicarbonyls from Dairy Related Carbohydrates with and without Nα-Acetyl-l-Lysine during Incubation at 40 and 50 °C.
Journal of agricultural and food chemistry (2019-05-16)
Wei Zhang, Mahesha M Poojary, Karsten Olsen, Colin A Ray, Marianne N Lund
ABSTRACT
α-Dicarbonyls are reactive intermediates formed during Maillard reactions and carbohydrate degradation. The formation of seven α-dicarbonyls was characterized in solutions containing dairy related carbohydrates (galactose, glucose, lactose, and galacto-oligosaccharides (GOS)) during incubations at 40 and 50 °C with and without Nα-acetyl-l-lysine at pH 6.8 for up to 2 months. The concentrations of α-dicarbonyls in samples of monosaccharides with Nα-acetyl-l-lysine were found to be 3-deoxyglucosone (3-DG) > 3-deoxygalactosone (3-DGal) > glyoxal > glucosone, galactosone > methylglyoxal > diacetyl. The presence of Nα-acetyl-l-lysine resulted in up to 100-fold higher concentrations of C6 α-dicarbonyls but lesser formation of glyoxal in the monosaccharide-containing models compared to what was observed in the absence of Nα-acetyl-l-lysine. Galactose incubated with Nα-acetyl-l-lysine generated the highest concentrations of 3-DGal (up to 130 μM), glyoxal (up to 100 μM), and methylglyoxal (up to 9 μM) compared to the other carbohydrates during incubation. Surprisingly, 3-DG (1500 μM) and 3-DGal (80 μM) were formed at levels of 2 orders of magnitude higher in solutions of GOS in the absence of Nα-acetyl-l-lysine as compared to the other carbohydrates at 40 °C, while GOS generated the lowest levels of glyoxal. GOS are widely used as an ingredient in various types of foods products, and it is therefore of importance to consider the risk of generating high levels of the reactive C6 α-dicarbonyl, 3-DG, in these types of products. This study contributes to the understanding of major α-dicarbonyl formation as affected by the presence of primary amines in GOS-, lactose-, and galactose-containing solutions under moderate heating in liquid foods.