MilliporeSigma
  • The Anti-amyloid Compound DO1 Decreases Plaque Pathology and Neuroinflammation-Related Expression Changes in 5xFAD Transgenic Mice.
The Anti-amyloid Compound DO1 Decreases Plaque Pathology and Neuroinflammation-Related Expression Changes in 5xFAD Transgenic Mice.
Cell chemical biology (2018-11-26)
Annett Boeddrich, Julius T Babila, Thomas Wiglenda, Lisa Diez, Manuela Jacob, Wilfried Nietfeld, Matthew R Huska, Christian Haenig, Nicole Groenke, Alexander Buntru, Eric Blanc, Jochen C Meier, Elisabetta Vannoni, Christian Erck, Beate Friedrich, Henrik Martens, Nancy Neuendorf, Sigrid Schnoegl, David P Wolfer, Maarten Loos, Dieter Beule, Miguel A Andrade-Navarro, Erich E Wanker
ABSTRACT
Self-propagating amyloid-β (Aβ) aggregates or seeds possibly drive pathogenesis of Alzheimer's disease (AD). Small molecules targeting such structures might act therapeutically in vivo. Here, a fluorescence polarization assay was established that enables the detection of compound effects on both seeded and spontaneous Aβ42 aggregation. In a focused screen of anti-amyloid compounds, we identified Disperse Orange 1 (DO1) ([4-((4-nitrophenyl)diazenyl)-N-phenylaniline]), a small molecule that potently delays both seeded and non-seeded Aβ42 polymerization at substoichiometric concentrations. Mechanistic studies revealed that DO1 disrupts preformed fibrillar assemblies of synthetic Aβ42 peptides and decreases the seeding activity of Aβ aggregates from brain extracts of AD transgenic mice. DO1 also reduced the size and abundance of diffuse Aβ plaques and decreased neuroinflammation-related gene expression changes in brains of 5xFAD transgenic mice. Finally, improved nesting behavior was observed upon treatment with the compound. Together, our evidence supports targeting of self-propagating Aβ structures with small molecules as a valid therapeutic strategy.