MilliporeSigma
  • Differential expression of TLRs 2, 4, 9, iNOS and TNF-α and arginase activity in peripheral blood monocytes from glucantime unresponsive and responsive patients with anthroponotic cutaneous leishmaniasis caused by Leishmania tropica.
Differential expression of TLRs 2, 4, 9, iNOS and TNF-α and arginase activity in peripheral blood monocytes from glucantime unresponsive and responsive patients with anthroponotic cutaneous leishmaniasis caused by Leishmania tropica.
Microbial pathogenesis (2018-11-07)
Razieh Tavakoli Oliaee, Iraj Sharifi, Ali Afgar, Abdollah Jafarzadeh, Amir Tavakoli Kareshk, Mehdi Bamorovat, Hamid Sharifi, Zahra Babaei, Amir Keyhani, Alireza Keyhani, Leili Abedi, Fatemeh Sharifi
ABSTRACT
Detection of the mechanism of host/parasite interactions in unresponsive forms of anthroponotic cutaneous leishmaniasis (ACL) caused by Leishmania tropica is helpful for immunotherapy and vaccine development. In the present study, the gene expression of toll-like receptors (TLRs), TNF-α, iNOS and also arginase (ARG) activity in monocytes from Glucantime unresponsive in comparison to responsive patients infected with L. tropica was investigated. In this case-control study, patients with unresponsive (n = 10) and responsive (n = 10) ACL were recruited. Gene expression of TLR2, TLR4, TLR9, TNF-α and iNOS was analyzed in L. tropica-exposed monocytes. The level of ARG activity in both isolated promastigotes and the lysates of monocytes was also determined. L. tropica-exposed monocytes represented higher expression of all three TLRs and TNF-α and lower expression of iNOS compared to unexposed ones in both groups of patients. Results revealed a significant down-regulation of TLR2 and TNF-α and up-regulation of TLR9 expression in unresponsive isolates in comparison to responsive ones. Besides, ARG level showed a significant increase in L. tropica-stimulated monocytes and cultured promastigotes from unresponsive isolates versus responsive ones. The decreased TLR2, TLR4, TNF-α and iNOS and the increased level of TLR9 expression in L. tropica-exposed monocytes from unresponsive isolates and also the increment in ARG activity in their promastigotes and monocytes, might possibly be involved in the severity of the disease and leading to Glucantime unresponsiveness.