MilliporeSigma
  • Thermotropic Liquid-Crystalline and Light-Emitting Properties of Poly(pyridinium) Salts Containing Various Diamine Connectors and Hydrophilic Macrocounterions.
Thermotropic Liquid-Crystalline and Light-Emitting Properties of Poly(pyridinium) Salts Containing Various Diamine Connectors and Hydrophilic Macrocounterions.
Polymers (2019-05-15)
Tae Soo Jo, Haesook Han, Pradip K Bhowmik, Benoît Heinrich, Bertrand Donnio
ABSTRACT
A set of poly(pyridinium) salts containing various diamine moieties, as molecular connectors, and poly(ethyleneglycol)-4-nonylphenyl-3-sulfopropyl ether, thereafter referred to as "Macroion", as the hydrophilic counterion, were prepared by metathesis reaction from the respective precursory tosylated poly(pyridinium)s in methanol. The structure of these ionic polymers was established by spectroscopy and chromatography techniques. The shape-persistent ionic poly(pyridinium) materials, inserting rigid or semi-rigid diamine spacers, display thermotropic liquid-crystalline properties from room-temperature up to their isotropization (in the temperature range around 160-200 °C). The nature of the LC phases is lamellar in both cases as identified by the combination of various complementary experimental techniques including DSC, POM and variable-temperature SAXS. The other polymers, inserting bulky or flexible spacers, only form room temperature viscous liquids. These new macromolecular systems can then be referred to as polymeric ionic liquid crystals (PILCs) and or polymeric ionic liquids (PILs). All the ionic polymers show excellent thermal stability, in the 260-330 °C temperature range as determined by TGA measurements, and a good solubility in common organic solvents as well as in water. Their optical properties were characterized in both solution and solid states by UV-Vis and photoluminescent spectroscopies. They emit blue or green light in both the states and exhibit a positive solvatochromic effect.