MilliporeSigma
  • 2-(2-Aminothiazol-4-yl)pyrrolidine-based tartrate diamides as potent, selective and orally bioavailable TACE inhibitors.
2-(2-Aminothiazol-4-yl)pyrrolidine-based tartrate diamides as potent, selective and orally bioavailable TACE inhibitors.
Bioorganic & medicinal chemistry letters (2011-04-05)
Chaoyang Dai, Dansu Li, Janeta Popovici-Muller, Lianyun Zhao, Vinay M Girijavallabhan, Kristin E Rosner, Brian J Lavey, Razia Rizvi, Bandarpalle B Shankar, Michael K C Wong, Zhuyan Guo, Peter Orth, Corey O Strickland, Jing Sun, Xiaoda Niu, Shiying Chen, Joseph A Kozlowski, Daniel J Lundell, John J Piwinski, Neng-Yang Shih, M Arshad Siddiqui
ABSTRACT
TNF-α converting enzyme (TACE) inhibitors are promising agents to treat inflammatory disorders and cancer. We have investigated novel tartrate diamide TACE inhibitors where the tartrate core binds to zinc in a unique tridentate fashion. Incorporating (R)-2-(2-N-alkylaminothiazol-4-yl)pyrrolidines into the left hand side amide of the tartrate scaffold led to the discovery of potent and selective TACE inhibitors, some of which exhibited good rat oral bioavailability.
Sigma-Aldrich
Cyclopropylboronic acid