MilliporeSigma
  • Pathologic Prognostic Factors of Pineal Parenchymal Tumor of Intermediate Differentiation.
Pathologic Prognostic Factors of Pineal Parenchymal Tumor of Intermediate Differentiation.
Applied immunohistochemistry & molecular morphology : AIMM (2017-08-12)
Debajyoti Chatterjee, Khushboo Lath, Navneet Singla, Narendra Kumar, Bishan D Radotra
ABSTRACT
Pineal parenchymal tumor of intermediate differentiation (PPTID) is an uncommon tumor of the pineal gland. Although this behaves as a grade II/III tumor, the exact clinical behavior is not well known. There is no well-established pathologic factor that can predict the behavior of PPTID. The aim of this study was to determine the pathologic prognostic factors in PPTID. All PPTID cases diagnosed between 2006 and 2016 were analyzed retrospectively. Immunohistochemistry for synaptophysin, neurofilament protein (NFP), glial fibrillar acid protein, NeuN, and Ki-67 were performed in all cases. Cases were classified arbitrarily into low grade (mitosis <4/10 hpf and Ki-67 <5%) and high grade (mitosis ≥4/10 hpf and Ki-67 ≥5%). Clinical details including follow-up information were retrieved from the patients' files. A total of 16 patients (6 low grade and 10 high grade) were included in this study. The age ranged from 2 to 55 years (average, 28.2) with a mild male preponderance (male:female, 1.67:1). All cases showed strong and diffuse positivity for synaptophysin. Focal NFP positivity was seen in 2 low-grade and 3 high-grade tumors. Only 2 cases showed focal NeuN positivity. Average Ki-67 index was 1.7% and 12.6% in low-grade and high-grade tumors, respectively. All patients with low-grade tumor were alive without recurrence. Among the patients with high-grade tumors, 2 had local recurrence, 1 had spinal metastasis, and 3 patients died. Mitosis and Ki-67 proliferation index are the most important pathologic prognostic factors in PPTID. NFP expression does not carry any prognostic significance.