MilliporeSigma
  • Fibrinogenolytic toxin from Indian monocled cobra (Naja kaouthia) venom.
Fibrinogenolytic toxin from Indian monocled cobra (Naja kaouthia) venom.
Journal of biosciences (2011-06-10)
C Chandra Sekhar, Dibakar Chakrabarty
ABSTRACT
A fibrinogenolytic toxin of molecular weight 6.5 kDa has been purified from the venom of Indian monocled cobra (Naja kaouthia) by repeated cation exchange chromatography on CM-sephadex C-50. The purified toxin did not show any phospholipase activity but was mildly hemolytic on human erythrocytes. This toxin, called Lahirin, cleaved fibrinogen in a dose- and time-dependent manner. The digestion process apparently started with the A alpha chain, and gradually other lower-molecular-weight chains were also cleaved to low-molecular-weight peptides. The fibrinolytic activity was completely lost after treatment with ethylene di-amine tetra acetic acid (EDTA). However, exposure to 100 degree C for 1 min or pre-treatment with phenyl methyl sulfonyl fluoride (PMSF) did not affect the fibrinolytic activity. Cleavage of di-sulphide bonds by beta-mercaptoethanol or unfolding the protein with 4 M urea caused complete loss of activity of pure Lahirin.
Sigma-Aldrich
CM Sephadex® C-50