MilliporeSigma
MORC proteins and epigenetic regulation.
Plant signaling & behavior (2012-10-18)
Zdravko J Lorković
ABSTRACT
Two recent studies in Arabidopsis implicated MORC proteins, which contain a GHKL ATPase domain, in transcriptional gene silencing. Here, these studies are compared and contrasted to discuss the roles of MORC proteins in epigenetic regulation. Although MORC proteins are likely to catalyze changes in chromatin structure in response to epigenetic signals, their precise mode of action and target site-specificity still remain unclear.
Sigma-Aldrich
ChIPAb+ Acetyl-Histone H3 - ChIP Validated Antibody and Primer Set, from rabbit
Sigma-Aldrich
ChIPAb+ Monomethyl-Histone H3 (Lys27) - ChIP Validated Antibody and Primer Set, serum, from rabbit