MilliporeSigma
  • Tumor-induced angiogenesis studied in confrontation cultures of multicellular tumor spheroids and embryoid bodies grown from pluripotent embryonic stem cells.
Tumor-induced angiogenesis studied in confrontation cultures of multicellular tumor spheroids and embryoid bodies grown from pluripotent embryonic stem cells.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2001-04-09)
M Wartenberg, F Dönmez, F C Ling, H Acker, J Hescheler, H Sauer
ABSTRACT
Tumor vascularization is the rate-limiting step for the progression of cancer. Differential steps of tumor-induced angiogenesis were studied by a novel in vitro confrontation culture of avascular multicellular prostate tumor spheroids and embryoid bodies grown from pluripotent embryonic stem (ES) cells. Vascularization in embryoid bodies started on day 5 of cell culture and was paralleled by down-regulation of hypoxia-inducible factor 1 alpha (HIF-1 alpha) and vascular endothelial growth factor (VEGF). In parallel, a dissipation of gradients in the pericellular oxygen pressure was observed as measured by O(2)-sensitive microelectrodes. After 24--48 h of confrontation culture, cells positive for platelet endothelial cell adhesion molecule (PECAM-1) became visible in the contact region between the embryoid body and the tumor spheroid and sprouted within the confrontation cultures during subsequent days. Tumor-induced angiogenesis resulted in growth stimulation of tumor spheroids, disappearance of central necrosis and a reduction of the pericellular oxygen pressure. Furthermore, tumor vascularization resulted in elevated levels of HIF-1 alpha, VEGF, heat shock protein 27 (HSP27), and P-glycoprotein. Tumor-induced angiogenesis may augment the oxygen consumption in tumors resulting in an increased expression of hypoxia-related, proangiogenic genes as well as of HSP27 and P-glycoprotein, which are involved in a multidrug resistance phenotype.