MilliporeSigma
  • Transient aggregation of ubiquitinated proteins during dendritic cell maturation.
Transient aggregation of ubiquitinated proteins during dendritic cell maturation.
Nature (2002-05-10)
Hugues Lelouard, Evelina Gatti, Fanny Cappello, Olivia Gresser, Voahirana Camosseto, Philippe Pierre
ABSTRACT
Dendritic cells (DCs) are antigen-presenting cells with the unique capacity to initiate primary immune responses. Dendritic cells have a remarkable pattern of differentiation (maturation) that exhibits highly specific mechanisms to control antigen presentation restricted by major histocompatibility complex (MHC). MHC class I molecules present to CD8(+) cytotoxic T cells peptides that are derived mostly from cytosolic proteins, which are ubiquitinated and then degraded by the proteasome. Here we show that on inflammatory stimulation, DCs accumulate newly synthesized ubiquitinated proteins in large cytosolic structures. These structures are similar to, but distinct from, aggresomes and inclusion bodies observed in many amyloid diseases. Notably, these dendritic cell aggresome-like induced structures (DALIS) are transient, require continuous protein synthesis and do not affect the ubiquitin-proteasome pathway. Our observations suggest the existence of an organized prioritization of protein degradation in stimulated DCs, which is probably important for regulating MHC class I presentation during maturation.