MilliporeSigma
  • Oleate activates SREBP-1 signaling activity in SCD1-deficient hepatocytes.
Oleate activates SREBP-1 signaling activity in SCD1-deficient hepatocytes.
American journal of physiology. Endocrinology and metabolism (2017-08-31)
Mohamed A Lounis, Karl-F Bergeron, Maggie S Burhans, James M Ntambi, Catherine Mounier
ABSTRACT
Stearoyl-CoA desaturase-1 (SCD1) is a key player in lipid metabolism. SCD1 catalyzes the synthesis of monounsaturated fatty acids (MUFA). MUFA are then incorporated into triacylglycerols and phospholipids. Previous studies have shown that Scd1 deficiency in mice induces metabolic changes in the liver characterized by a decrease in de novo lipogenesis and an increase in β-oxidation. Interestingly, Scd1-deficient mice show a decrease in the expression and maturation of the principal lipogenic transcription factor sterol receptor element binding protein-1 (SREBP-1). The mechanisms mediating this effect on de novo lipogenesis and β-oxidation have not been fully elucidated. We evaluated the role of SCD1 on de novo lipogenesis and β-oxidation in HepG2 cells. We also used Scd1-deficient mice and two strains of transgenic mice that produce either oleate (GLS5) or palmitoleate (GLS3) in a liver-specific manner. We demonstrate that the expression of β-oxidation markers increases in SCD1-deficient hepatocytes and suggest that this is due to an increase in cellular polyunsaturated fatty acid content. We also show that the changes in the level of SREBP-1 expression, for both the precursor and the mature forms, are mainly due to the lack of oleate in SCD1-deficient hepatocytes. Indeed, oleate treatment of cultured HepG2 cells or hepatic oleate production in chow-fed GLS5 mice can restore SREBP-1 expression and increase hepatic de novo lipogenesis. Finally, we show that oleate specifically increases SREBP-1 nuclear accumulation, suggesting a central role for oleate in SREBP-1 signaling activity.
Sigma-Aldrich
A939572, ≥98% (HPLC)