MilliporeSigma
A PIP Gets the plasmodium protein export pathway going.
Cell host & microbe (2012-02-22)
Rays H Y Jiang, Matthias Marti
ABSTRACT
Survival of blood stage malaria parasites requires extensive host cell remodeling, which is facilitated by secretion of parasite proteins via a dedicated protein export pathway. In a recent Cell paper, Bhattacharjee et al., (2012) describe PI(3)P binding as one of the first steps in targeting parasite proteins to the host cell.
Product Description
Avanti
18:1 PI(3,4,5)P3, 1,2-dioleoyl-sn-glycero-3-phospho-(1′-myo-inositol-3′,4′,5′-trisphosphate) (ammonium salt), powder
Avanti
06:0 PI(3,4,5)P3, 1,2-dihexanoyl-sn-glycero-3-phospho-(1′-myo-inositol-3′,4′,5′-trisphosphate) (ammonium salt), powder
Avanti
08:0 PI(3,4,5)P3, 1,2-dioctanoyl-sn-glycero-3-phospho-(1′-myo-inositol-3′,4′,5′-trisphosphate) (ammonium salt), powder