MilliporeSigma
  • Human cytomegalovirus infection downregulates vitamin-D receptor in mammalian cells.
Human cytomegalovirus infection downregulates vitamin-D receptor in mammalian cells.
The Journal of steroid biochemistry and molecular biology (2016-08-16)
Franz J J Rieder, Charlotte Gröschel, Marie-Theres Kastner, Karin Kosulin, Johannes Laengle, Rene Zadnikar, Rodrig Marculescu, Martina Schneider, Thomas Lion, Michael Bergmann, Enikö Kallay, Christoph Steininger
ABSTRACT
Vitamin D (VD) is essential for the human body and involved in a wide variety of critical physiological processes including bone, muscle, and cardiovascular health, as well as innate immunity and antimicrobial responses. Here, we elucidated the significance of the VD system in cytomegalovirus (CMV) infection, which is one of the most common opportunistic infections in immunocompromised or -suppressed patients. We found that expression of vitamin D receptor (VDR) was downregulated in CMV-infected cells within 12h [hrs] post infection [p.i.] to 12% relative to VDR expression in mock-infected fibroblasts and did not recover during the CMV replication cycle of 96h. None of the biologically active metabolites of VD, cholecalciferol, calcidiol, or calcitriol, inhibit CMV replication significantly in human fibroblasts. In a feedback loop, expression of CYP24A1 dropped to 3% by 12h p.i. and expression of CYP27B1 increased gradually during the replication cycle of CMV to 970% probably as a consequence of VDR inhibition. VDR expression was not downregulated during influenza virus or adenovirus replication. The potent synthetic vitamin D analog EB-1089 was not able to inhibit CMV replication or antagonize its effect on VDR expression. Only CMV replication, and none of the other viral pathogens evaluated, inhibited the vitamin D system in vitro. In view of the pleiotropism of VDR, CMV-mediated downregulation may have far-reaching virological, immunological, and clinical implications and thus warrant further evaluations in vitro and in vivo.
Sigma-Aldrich
Anti-Vitamin D Receptor antibody produced in rabbit, affinity isolated antibody