MilliporeSigma
  • Kinetics and Characterization of Degradation Products of Dihydralazine and Hydrochlorothiazide in Binary Mixture by HPLC-UV, LC-DAD and LC-MS Methods.
Kinetics and Characterization of Degradation Products of Dihydralazine and Hydrochlorothiazide in Binary Mixture by HPLC-UV, LC-DAD and LC-MS Methods.
Chromatographia (2018-09-18)
Anna Gumieniczek, Justyna Galeza, Tomasz Mroczek, Krzysztof Wojtanowski, Katarzyna Lipska, Rafał Pietras
ABSTRACT
Dihydralazine and hydrochlorothiazide were stored at high temperature and humidity, under UV/Vis light and different pH, as individual drugs and the mixture. Then, a sensitive and selective HPLC-UV method was developed for simultaneous determination of dihydralazine and hydrochlorothiazide in presence of their degradation products. Finally, the degradation products were characterized through LC-DAD and LC-MS methods. Dihydralazine was sensitive to high temperature and humidity, UV/Vis light and pH ≥ 7. At the same time, it was resistant to acidic conditions. Hydrochlorothiazide was sensitive to high temperature and humidity, UV/Vis light and changes in pH. Its highest level of degradation was observed in 1 M HCl. Degradation of the drugs was higher when they were stressed in the mixture. In the case of dihydralazine, the percentage degradation was 5-15 times higher. What is more, dihydralazine became sensitive to acidic conditions. Hydrochlorothiazide was shown to be more sensitive to UV/Vis light and pH > 4. Degradation of dihydralazine and hydrochlorothiazide followed first-order kinetics. The quickest degradation of dihydralazine was found to be in 1 M NaOH while of hydrochlorothiazide was in 1 M HCl (individual hydrochlorothiazide) or at pH 7-10 (hydrochlorothiazide in the mixture). A number of new degradation products were detected and some of them were identified by our LC-DAD and LC-MS methods. In the stressed individual samples, (phenylmethyl)hydrazine and 1,2,4-benzothiadiazine-7-sulfonamide 1,1-dioxide were observed for the first time. Interactions between dihydralazine and hydrochlorothiazide in the mixture were confirmed by additional degradation products, e.g., 2H-1,2,4-benzothiadiazine-7-sulfonamide 1,1,4-trioxide.
Sigma-Aldrich
Dihydralazine sulfate