MilliporeSigma
  • Fine mapping of five human skeletal muscle genes: alpha-tropomyosin, beta-tropomyosin, troponin-I slow-twitch, troponin-I fast-twitch, and troponin-C fast.
Fine mapping of five human skeletal muscle genes: alpha-tropomyosin, beta-tropomyosin, troponin-I slow-twitch, troponin-I fast-twitch, and troponin-C fast.
Biochemical and biophysical research communications (1997-01-13)
N Tiso, L Rampoldi, A Pallavicini, R Zimbello, D Pandolfo, G Valle, G Lanfranchi, G A Danieli
ABSTRACT
In this paper the chromosomal localization of the human skeletal muscle genes Troponin-I slow-twitch (TNNI1), Troponin-I fast-twitch (TNNI2), and Troponin-C fast (TNNC2) and the refinement of the position for alpha-Tropomyosin (TPM1) and beta-Tropomyosin (TPM2) are reported. By radiation hybrid mapping, TPM1 was assigned to chromosome 15q22.1, TPM2 to chromosome 9p13.2-p13.1, TNNI1 to chromosome 1q31.3, TNNI2 to chromosome 11p15.5, and TNNC2 to chromosome 20q12-q13.11. The genomic distribution of these genes is discussed, with particular emphasis on the cluster organization of the Troponin genes.
Sigma-Aldrich
Troponin1, His tagged human, recombinant, expressed in E. coli, ≥70% (SDS-PAGE), buffered aqueous glycerol solution