MilliporeSigma
  • Synthesis and vasodilatory activity of some amide derivatives of 6-(4-carboxymethyloxyphenyl)-4,5-dihydro-3(2H)-pyridazinone.
Synthesis and vasodilatory activity of some amide derivatives of 6-(4-carboxymethyloxyphenyl)-4,5-dihydro-3(2H)-pyridazinone.
European journal of medicinal chemistry (2009-07-11)
Ranju Bansal, Dinesh Kumar, Rosalia Carron, Carmen de la Calle
ABSTRACT
Synthesis and vasodilatory activity of some amide derivatives of 6-(4-carboxymethyloxyphenyl)-4,5-dihydro-3(2H)-pyridazinone are reported. An effect of substitution at 2-position of pyridazinone ring on vasodilatory potential has also been explored. The most active compound 6-[4-(2-oxo-2-pyrrolidin-1-yl-ethoxy)phenyl]-2-(4-fluorophenyl)-4,5-dihydropyridazin-3(2H)-one (11) exhibited vasodilating activity in nanomolar range (IC(50)=0.051 microM).
Sigma-Aldrich
2-Hydrazino-2-imidazoline hydrobromide, 98%