MilliporeSigma
  • Globally asynchronous sulphur isotope signals require re-definition of the Great Oxidation Event.
Globally asynchronous sulphur isotope signals require re-definition of the Great Oxidation Event.
Nature communications (2018-06-10)
Pascal Philippot, Janaína N Ávila, Bryan A Killingsworth, Svetlana Tessalina, Franck Baton, Tom Caquineau, Elodie Muller, Ernesto Pecoits, Pierre Cartigny, Stefan V Lalonde, Trevor R Ireland, Christophe Thomazo, Martin J van Kranendonk, Vincent Busigny
ABSTRACT
The Great Oxidation Event (GOE) has been defined as the time interval when sufficient atmospheric oxygen accumulated to prevent the generation and preservation of mass-independent fractionation of sulphur isotopes (MIF-S) in sedimentary rocks. Existing correlations suggest that the GOE was rapid and globally synchronous. Here we apply sulphur isotope analysis of diagenetic sulphides combined with U-Pb and Re-Os geochronology to document the sulphur cycle evolution in Western Australia spanning the GOE. Our data indicate that, from ~2.45 Gyr to beyond 2.31 Gyr, MIF-S was preserved in sulphides punctuated by several episodes of MIF-S disappearance. These results establish the MIF-S record as asynchronous between South Africa, North America and Australia, argue for regional-scale modulation of MIF-S memory effects due to oxidative weathering after the onset of the GOE, and suggest that the current paradigm of placing the GOE at 2.33-2.32 Ga based on the last occurrence of MIF-S in South Africa should be re-evaluated.
Sigma-Aldrich
1,1′-Carbonyl-di-(1,2,4-triazole), technical, ≥90% (T)