MilliporeSigma
  • Insulin-like growth factor binding protein-4 inhibits both basal and IGF-mediated chick pelvic cartilage growth in vitro.
Insulin-like growth factor binding protein-4 inhibits both basal and IGF-mediated chick pelvic cartilage growth in vitro.
Journal of bone and mineral research : the official journal of the American Society for Bone and Mineral Research (1993-04-01)
P M Schiltz, S Mohan, D J Baylink
ABSTRACT
This laboratory has purified a unique insulin-like growth factor binding protein (IGFBP-4) that was previously demonstrated to be inhibitory to bone cell proliferation. In this study, the hypothesis that IGFBP-4 is inhibitory to insulin-like growth factor (IGF) actions on cartilage was tested using the pelvic cartilages of 10-day-old chick embryos as an in vitro model system. Pelvic leaflets were incubated in serum-free medium for 18 h with effectors (BSA, IGF-I, IGF-II, IGFBP-4, or a combination of IGF and IGFBP-4). After the first 8 h, 1.5 microCi [3H]thymidine per well was added. Cartilage growth was assayed by TCA-insoluble [3H]thymidine incorporation into DNA. Additional experiments were conducted under similar conditions to assess the actions of the effectors on cartilage dry weight over a 72 h time period. In separate experiments, serum-free medium conditioned by chick pelvic cartilages for 72 h was assayed for IGF-II by radioreceptorassay, IGF-I by radioimmunoassay, and IGFBP by western ligand analysis. Exogenous IGF addition increased [3H]thymidine incorporation and dry weight of cartilages compared to controls. IGFBP-4 decreased both parameters in basal cartilage growth and also inhibited IGF-mediated cartilage growth. Pelvic cartilages secreted in vitro both IGF-I and IGF-II and a 32-34 kD IGFBP. In conclusion, the IGFs are stimulatory to cartilage growth in vitro and embryonic chick cartilage in vitro produces both IGF-I and II as well as an IGFBP.(ABSTRACT TRUNCATED AT 250 WORDS)
Sigma-Aldrich
IGF-Binding Protein 4 human, recombinant, expressed in (BTI-Tn-5B1-4) High-5 Insect Cells, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture