MilliporeSigma
  • Tyrosine kinase activity of EphA2 promotes its S897 phosphorylation and glioblastoma cell proliferation.
Tyrosine kinase activity of EphA2 promotes its S897 phosphorylation and glioblastoma cell proliferation.
Biochemical and biophysical research communications (2018-04-08)
Yuho Hamaoka, Manabu Negishi, Hironori Katoh
ABSTRACT
EphA2, a member of the Eph family of receptor tyrosine kinases, has been reported to promote tumor malignancy through phosphorylation of serine 897 (S897). Here, we found that overexpression of wild-type EphA2 induced S897 phosphorylation through ERK activation without growth factors or cytokines and promoted glioblastoma cell proliferation. However, overexpression of a kinase-inactive mutant of EphA2 failed to induce ERK activation, S897 phosphorylation, and promotion of glioblastoma cell proliferation. These data suggest that when overexpressed, EphA2 induces ERK activation through its tyrosine kinase activity, leading to S897 phosphorylation and promotion of glioblastoma cell proliferation. Our findings provide a new insight into how EphA2 mediates glioblastoma progression.
Sigma-Aldrich
2-Morpholinoethyl isocyanide, ≥98.0% (GC)