Skip to Content
MilliporeSigma
Z-Thr-OMe 98%

Z-Thr-OMe

Z-Thr-OMe 98%
Synonym(s):
N-Carbobenzyloxy-L-threonine methyl ester, Z-L-Threonine methyl ester
Linear Formula:
CH3CH(OH)CH(NHCO2CH2C6H5)CO2CH3
CAS Number:
Molecular Weight:
267.28

Products

Brand
Formula Weight
Melting Point (°C)
Purity
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Amino Acid Type
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing