Skip to Content
MilliporeSigma
(S)-(+)-2-Methylbutyric acid 98%

(S)-(+)-2-Methylbutyric acid

(S)-(+)-2-Methylbutyric acid 98%
Linear Formula:
C2H5CH(CH3)CO2H
CAS Number:
Molecular Weight:
102.13

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing