Skip to Content
MilliporeSigma
Prednisolone British Pharmacopoeia (BP) Assay Standard

Prednisolone

Prednisolone British Pharmacopoeia (BP) Assay Standard
Synonym(s):
1,4-Pregnadiene-11β,17α,21-triol-3,20-dione, 1-Dehydrocortisol, 1-Dehydrohydrocortisone, 11β,17α,21-Trihydroxy-1,4-pregnadiene-3,20-dione
Empirical Formula (Hill Notation):
C21H28O5
CAS Number:
Molecular Weight:
360.44
EC Number:

Products

Product Category
Brand
Biological Source
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Format
Research Area
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Sterility
Technique