Skip to Content
MilliporeSigma
PCB No 101 analytical standard

PCB No 101

PCB No 101 analytical standard
Synonym(s):
2,2′,4,5,5′-Pentachlorobiphenyl
Empirical Formula (Hill Notation):
C12H5Cl5
CAS Number:
Molecular Weight:
326.43
EC Number:

Products

Brand
Formula Weight
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing