Skip to Content
MilliporeSigma
N-(2-Hydroxyethyl)trifluoroacetamide 97%

N-(2-Hydroxyethyl)trifluoroacetamide

N-(2-Hydroxyethyl)trifluoroacetamide 97%
Synonym(s):
N-(Trifluoroacetyl)ethanolamine
Linear Formula:
CF3CONHCH2CH2OH
CAS Number:
Molecular Weight:
157.09

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Functional Group
Markush Class
Markush Group
Reagent Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing