Skip to Content
MilliporeSigma
Molybdenum(VI) oxide ACS reagent, ≥99.5%

Molybdenum(VI) oxide

Molybdenum(VI) oxide ACS reagent, ≥99.5%
Synonym(s):
Molybdenum trioxide, Molybdenum(VI) oxide
Linear Formula:
MoO3
CAS Number:
Molecular Weight:
143.94

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Application
Reagent Type
Core
Special Grade