
Methoxyacetyl chloride

Synonym(s):
Methoxyacetic acid chloride
Linear Formula:
CH3OCH2COCl
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing