
Mesna Related Compound A

Synonym(s):
2-(Acetylsulfanyl)ethanesulfonic acid potassium salt-KCl adduct
Empirical Formula (Hill Notation):
C4H7KO4S2 · KCl
Molecular Weight:
296.88
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing