Skip to Content
MilliporeSigma
L-Isoleucine-13C6,15N 98 atom % 15N, 98 atom % 13C, 95% (CP)

L-Isoleucine-13C6,15N

L-Isoleucine-13C6,15N 98 atom % 15N, 98 atom % 13C, 95% (CP)
Synonym(s):
(2S,3S)-2-Amino-3-methylpentanoic acid-13C6,15N, 13C and 15N Labeled L-isoleucine
Linear Formula:
13C2H513CH(13CH3)13CH(15NH2)13CO2H
CAS Number:
Molecular Weight:
138.12

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Isotope
Markush Class
Markush Group
Optical Isomer
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing