Skip to Content
MilliporeSigma
Latanoprost acid ≥95% (HPLC)

Latanoprost acid

Latanoprost acid ≥95% (HPLC)
Synonym(s):
(5Z,9α,11α,15R)-9,11,15-Trihydroxy-17-phenyl-18,19,20-trinor-prost-5-en-1-oic acid, (Z)-7-[(1R,2R,3R,5S)-3,5-Dihydroxy-2-[(3R)-3-hydroxy-5-phenylpentyl]cyclopentyl]hept-5-enoic acid, PhXA 85
Empirical Formula (Hill Notation):
C23H34O5
CAS Number:
Molecular Weight:
390.51

Products

Product Category
Brand
Color
Formula Weight
Gene Alias
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing