
Lacosamide for system suitability

Synonym(s):
Lacosamide
Empirical Formula (Hill Notation):
C13H18N2O3
CAS Number:
Molecular Weight:
250.29
Products
Formula Weight
Manufacturer
Format
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing