Skip to Content
MilliporeSigma
Hydroxyzine Dihydrochloride Pharmaceutical Secondary Standard; Certified Reference Material

Hydroxyzine dihydrochloride

Hydroxyzine Dihydrochloride Pharmaceutical Secondary Standard; Certified Reference Material
Synonym(s):
Hydroxyzine dihydrochloride
Empirical Formula (Hill Notation):
C21H27ClN2O2 · 2HCl
CAS Number:
Molecular Weight:
447.83
EC Number:

Products

Product Category
Brand
Formula Weight
Gene Alias
Physical Form
Purity
Agency
Format
Research Area
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique