Skip to Content
MilliporeSigma
Fmoc-Ile-OH ≥98.0% (T)

Fmoc-Ile-OH

Fmoc-Ile-OH ≥98.0% (T)
Synonym(s):
N-(9-Fluorenylmethoxycarbonyl)-L-isoleucine, Fmoc-L-isoleucine, Fmoc-Ile-OH
Empirical Formula (Hill Notation):
C21H23NO4
CAS Number:
Molecular Weight:
353.41

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Reaction Type
Reagent Type
Showing 1-2 of 2
Compare
Product Number
Description
Pricing