
Fluconazole impurity C

Synonym(s):
1,1′-(1,3-Phenylene)bis-1H-1,2,4-triazole
Empirical Formula (Hill Notation):
C10H8N6
CAS Number:
Molecular Weight:
212.21
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing