Skip to Content
MilliporeSigma
Ethanol-1-13C 95% in H2O, 99 atom % 13C

Ethanol-1-13C

Ethanol-1-13C 95% in H2O, 99 atom % 13C
Synonym(s):
Ethyl alcohol-1-13C
Linear Formula:
CH313CH2OH
CAS Number:
Molecular Weight:
47.06

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Quality Segment
Isotope
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing