Skip to Content
MilliporeSigma
Diphenhydramine hydrochloride British Pharmacopoeia (BP) Reference Standard

Diphenhydramine hydrochloride

Diphenhydramine hydrochloride British Pharmacopoeia (BP) Reference Standard
Synonym(s):
2-Diphenylmethoxy-N,N-dimethylethylamine hydrochloride, N-(2-Diphenylmethoxyethyl)-N,N-dimethylamine hydrochloride
Empirical Formula (Hill Notation):
C17H21NO · HCl
CAS Number:
Molecular Weight:
291.82
EC Number:

Products

Product Category
Formula Weight
Gene Alias
Manufacturer
Physical Form
Purity
Agency
Format
Research Area
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique