Skip to Content
MilliporeSigma
(+)-Borneol analytical standard

(+)-Borneol

(+)-Borneol analytical standard
Synonym(s):
endo-(1R)-1,7,7-Trimethylbicyclo[2.2.1]heptan-2-ol
Empirical Formula (Hill Notation):
C10H18O
CAS Number:
Molecular Weight:
154.25
EC Number:

Products

Product Category
Brand
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing