Skip to Content
MilliporeSigma
vibrant-m

Cyclizine hydrochloride

vibrant-m
Synonym(s):
1-Benzhydryl-4-methylpiperazine hydrochloride, Marezine hydrochloride
Empirical Formula (Hill Notation):
C18H22N2 · HCl
CAS Number:
Molecular Weight:
302.84

Products

Formula Weight
Manufacturer
Physical Form
Format
Application
API Family
Reference Material Type
Special Grade