
Cefradine for peak identification

Synonym(s):
Cephradine, Cefradin
Empirical Formula (Hill Notation):
C16H19N3O4S
CAS Number:
Molecular Weight:
349.40
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing