
Carbamazepine impurity A

Synonym(s):
10,11-Dihydrocarbamazepine, 10,11-Dihydro-5H-dibenz[b,f]azepine-5-carboxamide
Empirical Formula (Hill Notation):
C15H14N2O
CAS Number:
Molecular Weight:
238.28
Products
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing