Skip to Content
MilliporeSigma
Butyl butyrate natural, ≥98%, FCC, FG

Butyl butyrate

Butyl butyrate natural, ≥98%, FCC, FG
Linear Formula:
CH3CH2CH2COO(CH2)3CH3
CAS Number:
Molecular Weight:
144.21
EC Number:

Products

Product Category
Brand
Sustainability
Biological Source
Boiling Point (°C)
Feature
Formula Weight
Physical Form
Purity
Agency
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-3 of 3
Compare
Product Number
Description
Pricing