Skip to Content
MilliporeSigma
Bromothymol Blue sodium salt powder

Bromothymol blue sodium salt

Bromothymol Blue sodium salt powder
Synonym(s):
Bromothymol blue sodium salt, 3′,3′′-dibromothymolsulfonephthalein sodium salt
Empirical Formula (Hill Notation):
C27H27Br2NaO5S
CAS Number:
Molecular Weight:
646.36
EC Number:

Products

Product Category
Brand
Formula Weight
pH Value
Physical Form
Quality Segment
Application
Special Grade
Technique
Showing 1-3 of 3