Skip to Content
MilliporeSigma
Bergenin ≥95% (LC/MS-ELSD)

Bergenin

Bergenin ≥95% (LC/MS-ELSD)
Synonym(s):
Ardisic acid B, Cuscutin, Vakerin
Empirical Formula (Hill Notation):
C14H16O9
CAS Number:
Molecular Weight:
328.27

Products

Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Research Area
Application
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing