Skip to Content
MilliporeSigma
Bacitracin Pharmaceutical Secondary Standard; Certified Reference Material

Bacitracin

Bacitracin Pharmaceutical Secondary Standard; Certified Reference Material
Synonym(s):
Bacitracin
Empirical Formula (Hill Notation):
C66H103N17O16S
CAS Number:
Molecular Weight:
1422.69
EC Number:

Products

Product Category
Brand
Biological Source
Formula Weight
Melting Point (°C)
Physical Form
Activity Spectrum
Agency
Format
Quality Segment
Application
Mode of Action
Package Size
API Family
Reference Material Type
Special Grade
Technique