
Aurapten

Synonym(s):
Auraptene, 7-Geranyloxycoumarin, 7-[[(2E)-3,7-dimethyl-2,6-octadien-1-yl]oxy]-2H-1-benzopyran-2-one, Aurapten
Empirical Formula (Hill Notation):
C19H22O3
CAS Number:
Molecular Weight:
298.38
Products
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing